Assay Details
| Gene Symbol: ARFGEF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020340.4 | NP_065073.3 | 25 - 26 | 4367 | 69 | |
| GenBank mRNA | AB033070.2 | - | 16 - 17 | 3281 | 69 | |
| AB252196.1 | - | 25 - 26 | 4367 | 69 | ||
| AF413080.1 | - | 14 - 15 | 2975 | 69 | ||
| BC137174.1 | - | 23 - 24 | 4002 | 9021171 | 69 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001029.hg.1
Set Membership: |
|

ARFGEF3
Genomic Map