Assay Details
| Gene Symbol: ARFGEF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020340.4 | NP_065073.3 | 2 - 3 | 313 | 86 | |
| GenBank mRNA | AB252196.1 | - | 2 - 3 | 313 | 86 | |
| BF943924.1 | - | 2 - 3 | 86 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001029.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06012040.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06012040.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06012040.hg.1 | TC06001029.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06012041.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06012041.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06012041.hg.1 | TC06001029.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06012039.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06012039.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06012039.hg.1 | TC06001029.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009606.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600165830.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600165830.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600165830.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600165830.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600165830.hg.1 | TC0600009606.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600165833.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600165833.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600165833.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600165833.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600165833.hg.1 | TC0600009606.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600165831.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600165831.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600165831.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600165831.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600165831.hg.1 | TC0600009606.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600165832.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600165832.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600165832.hg.1 | ENST00000251691.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600165832.hg.1 | NM_020340.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600165832.hg.1 | TC0600009606.hg.1 | Clariom D TC |

ARFGEF3
Genomic Map