| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010526.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010526.hg.1 |
ENST00000443756.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010526.hg.1 |
ENST00000545132.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010526.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010526.hg.1 |
TC01000645.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010525.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010525.hg.1 |
ENST00000443756.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010525.hg.1 |
ENST00000545132.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010525.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010525.hg.1 |
TC01000645.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010527.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010527.hg.1 |
ENST00000443756.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010527.hg.1 |
ENST00000545132.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010527.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010527.hg.1 |
TC01000645.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100159332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100159332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100159332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100159332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100159332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100159332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100159332.hg.1 |
TC0100008332.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100159331.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100159331.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100159331.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100159331.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100159331.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100159331.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100159331.hg.1 |
TC0100008332.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100159333.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100159333.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100159333.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100159333.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100159333.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100159333.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100159333.hg.1 |
TC0100008332.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000294353.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
ENST00000545132.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008332.hg.1 |
NM_024646.2 |
Refseq |