| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038371.hg.1 |
ENST00000291416.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038371.hg.1 |
ENST00000373430.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038371.hg.1 |
ENST00000373432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038371.hg.1 |
ENST00000543586.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038371.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038371.hg.1 |
TC01002481.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038366.hg.1 |
ENST00000291416.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038366.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038366.hg.1 |
TC01002481.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038369.hg.1 |
ENST00000291416.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038369.hg.1 |
ENST00000543586.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038369.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038369.hg.1 |
TC01002481.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038367.hg.1 |
ENST00000291416.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038367.hg.1 |
ENST00000543586.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038367.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038367.hg.1 |
TC01002481.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000485148.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000485148.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000485148.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000485148.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000485148.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000486583.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000486583.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000486583.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000486583.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000543586.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000543586.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000543586.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000543586.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000543586.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
ENST00000543586.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100201517.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201517.hg.1 |
ENST00000543586.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201517.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100201517.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201517.hg.1 |
ENST00000543586.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201517.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100201517.hg.1 |
TC0100013647.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000485148.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000485148.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000485148.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000485148.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000485148.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000486583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000486583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000486583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000486583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000543586.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000543586.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000543586.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000543586.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000543586.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
ENST00000543586.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013647.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100201516.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201516.hg.1 |
ENST00000543586.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201516.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100201516.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201516.hg.1 |
ENST00000543586.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201516.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100201516.hg.1 |
TC0100013647.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100201515.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201515.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100201515.hg.1 |
ENST00000291416.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100201515.hg.1 |
NM_018207.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100201515.hg.1 |
TC0100013647.hg.1 |
Clariom D TC |