| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000361584.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000361584.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000361584.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000361584.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000361584.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000361584.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000361584.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000361584.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000361584.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000411995.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000411995.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000411995.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000411995.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000411995.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000411995.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000411995.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000411995.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000411995.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000411995.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000469416.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000469416.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000469416.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000469416.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000469416.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000469416.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000469416.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000469416.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000469416.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000469416.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000469416.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000484445.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000484445.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000484445.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000484445.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000484445.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000484445.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000484445.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
ENST00000484445.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
NM_023070.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
NM_023070.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
NM_023070.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
NM_023070.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
NM_023070.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
NM_023070.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
NM_023070.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
NM_023070.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007958.hg.1 |
NM_023070.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000361584.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000361584.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000361584.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000361584.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000361584.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000361584.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000361584.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000361584.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000361584.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000411995.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000411995.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000411995.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000411995.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000411995.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000411995.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000411995.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000411995.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000411995.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000411995.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000469416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000469416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000469416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000469416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000469416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000469416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000469416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000469416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000469416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000469416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000469416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000484445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000484445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000484445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000484445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000484445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000484445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000484445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
ENST00000484445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
NM_023070.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
NM_023070.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
NM_023070.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
NM_023070.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
NM_023070.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
NM_023070.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
NM_023070.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
NM_023070.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007958.hg.1 |
NM_023070.2 |
Refseq |