Assay Details
| Gene Symbol: ZFP69B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023070.2 | NP_075558.2 | 1 - 2 | 429 | 69 | |
| XM_005271136.1 | XP_005271193.1 | 2 - 3 | 512 | 69 | ||
| XM_005271137.2 | XP_005271194.1 | 2 - 3 | 512 | 69 | ||
| XM_017002147.1 | XP_016857636.1 | 2 - 3 | 462 | 69 | ||
| GenBank mRNA | AK091612.1 | - | 2 - 3 | 512 | 69 | |
| BC017498.1 | - | 1 - 2 | 50 | 4576469 | 69 | |
| CN256362.1 | - | 2 - 3 | 514 | 69 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000511.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK091612.1,BC017498.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01007973.hg.1 | ENST00000361584.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007973.hg.1 | ENST00000411995.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007973.hg.1 | ENST00000431552.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007973.hg.1 | ENST00000484445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007973.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007973.hg.1 | TC01000511.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007975.hg.1 | ENST00000361584.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007975.hg.1 | ENST00000411995.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007975.hg.1 | ENST00000431552.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007975.hg.1 | ENST00000484445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007975.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007975.hg.1 | TC01000511.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007974.hg.1 | ENST00000411995.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007974.hg.1 | ENST00000431552.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007974.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007974.hg.1 | TC01000511.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007972.hg.1 | ENST00000361584.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007972.hg.1 | ENST00000411995.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007972.hg.1 | ENST00000431552.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007972.hg.1 | ENST00000484445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007972.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007972.hg.1 | TC01000511.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100156017.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156017.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156017.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156017.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156017.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100156017.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156017.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156017.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156017.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156017.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100156017.hg.1 | TC0100007958.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100156020.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156020.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156020.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156020.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156020.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100156020.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156020.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156020.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156020.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156020.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100156020.hg.1 | TC0100007958.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100156016.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156016.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156016.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156016.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156016.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100156016.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156016.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156016.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156016.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156016.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100156016.hg.1 | TC0100007958.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100156018.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156018.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156018.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156018.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156018.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100156018.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156018.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156018.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156018.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156018.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100156018.hg.1 | TC0100007958.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100156019.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156019.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156019.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156019.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100156019.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156019.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156019.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156019.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100156019.hg.1 | TC0100007958.hg.1 | Clariom D TC |

ZFP69B
Genomic Map