Assay Details
| Gene Symbol: SRSF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK095112.1 | - | 1 - 2 | 3824 | 96 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000577267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000577267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000577267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000577267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000577267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000577267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000577267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000577267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000577267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000577267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000577267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000577267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000582096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000582096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000582096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000582096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000582096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000582096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000582096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000582096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000582096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000577267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000577267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000577267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000577267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000577267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000577267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000577267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000577267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000577267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000577267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000577267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000577267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000582096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000582096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000582096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000582096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000582096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000582096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000582096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000582096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000582096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012436.hg.1 | ENST00000585065.1 | Ensembl |

SRSF1
Genomic Map