| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
ENST00000562742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
ENST00000562742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
ENST00000562742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
ENST00000563754.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
ENST00000563754.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
ENST00000563754.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
ENST00000563754.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
ENST00000563754.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
ENST00000563754.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
NR_110918.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
NR_110918.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
NR_110918.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
NR_110918.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
NR_110918.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010563.hg.1 |
NR_110918.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600181160.hg.1 |
ENST00000562742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181160.hg.1 |
ENST00000563754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181160.hg.1 |
NR_110918.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600181160.hg.1 |
TC1600010563.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600181160.hg.1 |
ENST00000562742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181160.hg.1 |
ENST00000563754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181160.hg.1 |
NR_110918.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600181159.hg.1 |
ENST00000563754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181159.hg.1 |
NR_110918.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600181159.hg.1 |
TC1600010563.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600181159.hg.1 |
ENST00000563754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181159.hg.1 |
NR_110918.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
ENST00000562742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
ENST00000562742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
ENST00000562742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
ENST00000563754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
ENST00000563754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
ENST00000563754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
ENST00000563754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
ENST00000563754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
ENST00000563754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
NR_110918.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
NR_110918.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
NR_110918.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
NR_110918.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
NR_110918.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010563.hg.1 |
NR_110918.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600181162.hg.1 |
ENST00000562742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181162.hg.1 |
ENST00000563754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181162.hg.1 |
NR_110918.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600181162.hg.1 |
TC1600010563.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600181162.hg.1 |
ENST00000562742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181162.hg.1 |
ENST00000563754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600181162.hg.1 |
NR_110918.1 |
Refseq |