Assay Details
| Gene Symbol: ZC3H4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015168.1 | NP_055983.1 | 7 - 8 | 1003 | 87 | |
| XM_005258676.3 | XP_005258733.1 | 6 - 7 | 2278 | 87 | ||
| XM_005258677.4 | XP_005258734.1 | 7 - 8 | 1129 | 87 | ||
| XM_005258678.2 | XP_005258735.1 | 6 - 7 | 886 | 87 | ||
| XM_006723113.3 | XP_006723176.1 | 7 - 8 | 2693 | 87 | ||
| XM_011526668.2 | XP_011524970.1 | 7 - 8 | 905 | 87 | ||
| XM_011526669.2 | XP_011524971.1 | 6 - 7 | 1033 | 87 | ||
| XM_011526670.2 | XP_011524972.1 | 6 - 7 | 1629 | 87 | ||
| XM_017026529.1 | XP_016882018.1 | 6 - 7 | 2085 | 87 | ||
| XM_017026530.1 | XP_016882019.1 | 7 - 8 | 2693 | 87 | ||
| XM_017026531.1 | XP_016882020.1 | 5 - 6 | 784 | 87 | ||
| GenBank mRNA | AB028987.2 | - | 7 - 8 | 1003 | 87 | |
| AK125279.1 | - | 5 - 6 | 731 | 87 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001660.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19022567.hg.1 | ENST00000253048.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022567.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022567.hg.1 | TC19001660.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022566.hg.1 | ENST00000253048.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022566.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022566.hg.1 | TC19001660.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000595341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000595341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000595341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000597069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000597069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000597069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000597069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000597069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000597069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022568.hg.1 | ENST00000253048.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022568.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022568.hg.1 | TC19001660.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900191781.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900191781.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900191781.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900191781.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900191781.hg.1 | TC1900011014.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900191781.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900191781.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900191781.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900191781.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000595341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000595341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000595341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000597069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000597069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000597069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000597069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000597069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000597069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900191783.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900191783.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900191783.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900191783.hg.1 | TC1900011014.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900191783.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900191783.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900191783.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900191780.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900191780.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900191780.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900191780.hg.1 | TC1900011014.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900191780.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900191780.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900191780.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900191782.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900191782.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900191782.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900191782.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900191782.hg.1 | TC1900011014.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900191782.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900191782.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900191782.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900191782.hg.1 | NM_015168.1 | Refseq |

ZC3H4
Genomic Map