Assay Details
| Gene Symbol: VPS33A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022916.4 | NP_075067.2 | 11 - 12 | 1556 | 68 | |
| GenBank mRNA | AF439857.1 | - | 11 - 12 | 1467 | 68 | |
| AK026048.1 | - | 5 - 6 | 677 | 68 | ||
| AK026840.1 | - | 11 - 12 | 1466 | 68 | ||
| AK315545.1 | - | 11 - 12 | 1494 | 68 | ||
| BC016617.1 | - | 11 - 12 | 1464 | 3855313 | 68 | |
More Information
Set Membership: |
|

VPS33A
Genomic Map