Assay Details
| Gene Symbol: HSD3B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000198.3 | NP_000189.1 | 1 - 2 | 82 | 77 | |
| GenBank mRNA | AK222997.1 | - | 1 - 2 | 80 | 77 | |
| AK313413.1 | - | 1 - 2 | 52 | 77 | ||
| AV705363.1 | - | 1 - 2 | 118 | 77 | ||
| BC038419.1 | - | 1 - 2 | 57 | 5167092 | 77 | |
| CF596786.1 | - | 1 - 2 | 56 | 77 | ||
| M67466.1 | - | 1 - 2 | 43 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001039.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK313413.1,BC038419.1,M67466.1,NM_000198.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000433745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000433745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000433745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000433745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000433745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000433745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000443865.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000443865.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000443865.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000443865.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000443865.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000443865.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000448448.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000448448.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000448448.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000471656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000471656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000471656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000471656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000471656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016467.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016467.hg.1 | ENST00000471656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016467.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016467.hg.1 | TC01001039.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016469.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016469.hg.1 | ENST00000471656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016469.hg.1 | ENST00000543831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016469.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016469.hg.1 | NM_001166120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016469.hg.1 | TC01001039.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009617.hg.1 | ENST00000511509.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009617.hg.1 | ENST00000511509.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009617.hg.1 | ENST00000511509.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009617.hg.1 | NR_033781.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009617.hg.1 | NR_033781.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009617.hg.1 | NR_033781.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009617.hg.1 | NR_033781.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009617.hg.1 | NR_033781.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009617.hg.1 | NR_033781.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009617.hg.1 | NR_033781.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009617.hg.1 | ENST00000511509.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009617.hg.1 | ENST00000511509.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009617.hg.1 | ENST00000511509.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009617.hg.1 | NR_033781.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009617.hg.1 | NR_033781.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009617.hg.1 | NR_033781.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009617.hg.1 | NR_033781.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009617.hg.1 | NR_033781.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009617.hg.1 | NR_033781.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009617.hg.1 | NR_033781.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169164.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169164.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169164.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169164.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169164.hg.1 | TC0100009611.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100169169.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169169.hg.1 | ENST00000433745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169169.hg.1 | ENST00000443865.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169169.hg.1 | ENST00000471656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169169.hg.1 | ENST00000543831.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169169.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169169.hg.1 | NM_001166120.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169169.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169169.hg.1 | ENST00000433745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169169.hg.1 | ENST00000443865.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169169.hg.1 | ENST00000471656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169169.hg.1 | ENST00000543831.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169169.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169169.hg.1 | NM_001166120.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169169.hg.1 | TC0100009611.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100169170.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169170.hg.1 | ENST00000433745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169170.hg.1 | ENST00000443865.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169170.hg.1 | ENST00000471656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169170.hg.1 | ENST00000543831.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169170.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169170.hg.1 | NM_001166120.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169170.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169170.hg.1 | ENST00000433745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169170.hg.1 | ENST00000443865.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169170.hg.1 | ENST00000471656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169170.hg.1 | ENST00000543831.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169170.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169170.hg.1 | NM_001166120.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169170.hg.1 | TC0100009611.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100169167.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169167.hg.1 | ENST00000443865.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169167.hg.1 | ENST00000471656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169167.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169167.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169167.hg.1 | ENST00000443865.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169167.hg.1 | ENST00000471656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169167.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169167.hg.1 | TC0100009611.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000433745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000433745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000433745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000433745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000433745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000433745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000443865.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000443865.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000443865.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000443865.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000443865.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000443865.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000448448.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000448448.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000448448.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000471656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000471656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000471656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000471656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000471656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_000198.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_001166120.1 | Refseq |

HSD3B2
Genomic Map