Assay Details
| Gene Symbol: GML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002066.2 | NP_002057.1 | 2 - 3 | 169 | 108 | |
| GenBank mRNA | BC074930.2 | - | 1 - 2 | 97 | 30915488 | 108 |
| BC126336.1 | - | 2 - 3 | 158 | 8992052 | 108 | |
| BC126338.1 | - | 2 - 3 | 158 | 8992054 | 108 | |
| D84290.1 | - | 2 - 3 | 168 | 108 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC074930.2, BC126336.1, BC126338.1, D84290.1, NM_002066.2 transcripts.
Set Membership: |
|

GML
Genomic Map