| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21003643.hg.1 |
ENST00000284885.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21003643.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21003643.hg.1 |
TC21000305.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21003644.hg.1 |
ENST00000284885.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21003644.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21003644.hg.1 |
TC21000305.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000422787.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000422787.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000422787.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000422787.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000422787.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000422787.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000422787.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000422787.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000474775.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000474775.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
ENST00000474775.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100152034.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100152034.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100152034.hg.1 |
TC2100007676.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100152034.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100152034.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100152035.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100152035.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100152035.hg.1 |
TC2100007676.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100152035.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100152035.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000284885.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000422787.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000422787.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000422787.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000422787.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000422787.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000422787.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000422787.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000422787.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000474775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000474775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
ENST00000474775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007676.hg.1 |
NM_002772.2 |
Refseq |