| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18005646.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18005646.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18005646.hg.1 |
TC18000542.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000587399.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000587399.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000587399.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000587399.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000587842.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000587842.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000587842.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000587842.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000589204.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000589204.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000589204.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000590642.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000590642.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000590662.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000590662.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000590662.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
ENST00000590662.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000587399.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000587399.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000587399.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000587399.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000587842.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000587842.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000587842.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000587842.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000589204.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000589204.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000589204.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000590642.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000590642.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000590662.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000590662.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000590662.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
ENST00000590662.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008804.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800160606.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800160606.hg.1 |
NM_052947.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800160606.hg.1 |
TC1800008804.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800160606.hg.1 |
ENST00000361673.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800160606.hg.1 |
NM_052947.3 |
Refseq |