Assay Details
| Gene Symbol: SEMA3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006080.2 | NP_006071.1 | 13 - 14 | 1811 | 76 | |
| XM_005250110.3 | XP_005250167.1 | 16 - 17 | 2006 | 76 | ||
| XM_005250111.4 | XP_005250168.1 | 15 - 16 | 1846 | 76 | ||
| XM_006715839.3 | XP_006715902.1 | 14 - 15 | 1749 | 76 | ||
| XM_011515734.2 | XP_011514036.1 | 16 - 17 | 1831 | 76 | ||
| XM_017011673.1 | XP_016867162.1 | 14 - 15 | 1666 | 76 | ||
| GenBank mRNA | AK289954.1 | - | 13 - 14 | 1904 | 76 | |
| BC111416.1 | - | 13 - 14 | 1510 | 40034963 | 76 | |
| L26081.1 | - | 13 - 14 | 1511 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001568.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
neural crest cell migration
neuron migration
regulation of heart rate
apoptotic process
axon guidance
axonal fasciculation
negative regulation of epithelial cell migration
facial nerve structural organization
trigeminal nerve structural organization
nerve development
olfactory bulb development
branchiomotor neuron axon guidance
gonadotrophin-releasing hormone neuronal migration to the hypothalamus
ventral trunk neural crest cell migration
sympathetic nervous system development
dendrite morphogenesis
regulation of axon extension involved in axon guidance
negative regulation of axon extension involved in axon guidance
axon extension involved in axon guidance
sensory system development
negative chemotaxis
axonogenesis involved in innervation
dichotomous subdivision of terminal units involved in salivary gland branching
sympathetic ganglion development
trigeminal ganglion development
semaphorin-plexin signaling pathway
sympathetic neuron projection extension
sympathetic neuron projection guidance
neural crest cell migration involved in autonomic nervous system development
semaphorin-plexin signaling pathway involved in neuron projection guidance
semaphorin-plexin signaling pathway involved in axon guidance
neural crest cell migration involved in sympathetic nervous system development
facioacoustic ganglion development
positive regulation of male gonad development
positive regulation of neuron migration
semaphorin receptor binding
neuropilin binding
chemorepellent activity
neural crest cell migration
neuron migration
regulation of heart rate
apoptotic process
axon guidance
axonal fasciculation
negative regulation of epithelial cell migration
facial nerve structural organization
trigeminal nerve structural organization
nerve development
olfactory bulb development
branchiomotor neuron axon guidance
gonadotrophin-releasing hormone neuronal migration to the hypothalamus
ventral trunk neural crest cell migration
sympathetic nervous system development
dendrite morphogenesis
regulation of axon extension involved in axon guidance
negative regulation of axon extension involved in axon guidance
axon extension involved in axon guidance
sensory system development
negative chemotaxis
axonogenesis involved in innervation
dichotomous subdivision of terminal units involved in salivary gland branching
sympathetic ganglion development
trigeminal ganglion development
semaphorin-plexin signaling pathway
sympathetic neuron projection extension
sympathetic neuron projection guidance
neural crest cell migration involved in autonomic nervous system development
semaphorin-plexin signaling pathway involved in neuron projection guidance
semaphorin-plexin signaling pathway involved in axon guidance
neural crest cell migration involved in sympathetic nervous system development
facioacoustic ganglion development
positive regulation of male gonad development
positive regulation of neuron migration
semaphorin receptor binding
neuropilin binding
chemorepellent activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07023673.hg.1 | ENST00000265362.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07023673.hg.1 | ENST00000436949.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07023673.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07023673.hg.1 | TC07001568.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000420047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000420047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000420047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000420047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000420047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201033.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201033.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201033.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201033.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201033.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201033.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201033.hg.1 | TC0700013578.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000265362.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000420047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000420047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000420047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000420047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000420047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | ENST00000436949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | NM_006080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013578.hg.1 | NM_006080.2 | Refseq |

SEMA3A
Genomic Map