| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000309244.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000309244.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000309244.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000309244.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000309244.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000309244.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000309244.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000309244.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000354232.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000354232.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000354232.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000354232.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000354232.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000354232.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000354232.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000354232.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000354232.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000593992.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000593992.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000593992.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000593992.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000593992.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000594361.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000594361.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000594361.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000596253.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000596253.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000596253.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000596253.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
ENST00000596253.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
NM_001014763.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
NM_001014763.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
NM_001014763.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
NM_001014763.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
NM_001014763.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
NM_001014763.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
NM_001014763.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
NM_001014763.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
NM_001985.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
NM_001985.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
NM_001985.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
NM_001985.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
NM_001985.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
NM_001985.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
NM_001985.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012016.hg.1 |
NM_001985.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024115.hg.1 |
ENST00000309244.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024115.hg.1 |
ENST00000354232.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024115.hg.1 |
NM_001014763.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024115.hg.1 |
NM_001985.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024115.hg.1 |
TC19001774.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024118.hg.1 |
ENST00000354232.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024118.hg.1 |
NM_001014763.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024118.hg.1 |
TC19001774.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000309244.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000309244.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000309244.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000309244.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000309244.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000309244.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000309244.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000309244.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000354232.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000354232.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000354232.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000354232.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000354232.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000354232.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000354232.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000354232.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000354232.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000593992.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000593992.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000593992.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000593992.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000593992.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000594361.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000594361.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000594361.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000596253.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000596253.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000596253.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000596253.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
ENST00000596253.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
NM_001014763.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
NM_001014763.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
NM_001014763.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
NM_001014763.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
NM_001014763.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
NM_001014763.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
NM_001014763.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
NM_001014763.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
NM_001985.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
NM_001985.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
NM_001985.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
NM_001985.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
NM_001985.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
NM_001985.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
NM_001985.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012016.hg.1 |
NM_001985.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900207524.hg.1 |
ENST00000354232.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207524.hg.1 |
NM_001014763.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900207524.hg.1 |
TC1900012016.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900207524.hg.1 |
ENST00000354232.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207524.hg.1 |
NM_001014763.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900207521.hg.1 |
ENST00000309244.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207521.hg.1 |
ENST00000354232.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207521.hg.1 |
ENST00000593992.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207521.hg.1 |
ENST00000596253.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207521.hg.1 |
NM_001014763.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900207521.hg.1 |
NM_001985.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900207521.hg.1 |
TC1900012016.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900207521.hg.1 |
ENST00000309244.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207521.hg.1 |
ENST00000354232.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207521.hg.1 |
ENST00000593992.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207521.hg.1 |
ENST00000596253.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207521.hg.1 |
NM_001014763.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900207521.hg.1 |
NM_001985.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900207525.hg.1 |
ENST00000354232.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207525.hg.1 |
NM_001014763.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900207525.hg.1 |
TC1900012016.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900207525.hg.1 |
ENST00000354232.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207525.hg.1 |
NM_001014763.1 |
Refseq |