Assay Details
| Gene Symbol: MYL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021223.2 | NP_067046.1 | 3 - 4 | 306 | 74 | |
| XM_005249817.4 | XP_005249874.1 | 4 - 5 | 449 | 74 | ||
| XM_011515463.2 | XP_011513765.1 | 4 - 5 | 450 | 74 | ||
| XM_011515464.2 | XP_011513766.1 | 4 - 5 | 380 | 74 | ||
| XM_011515465.2 | XP_011513767.1 | 4 - 5 | 380 | 74 | ||
| XM_017012478.1 | XP_016867967.1 | 4 - 5 | 379 | 74 | ||
| XM_017012479.1 | XP_016867968.1 | 4 - 5 | 387 | 74 | ||
| GenBank mRNA | AK311869.1 | - | 4 - 5 | 321 | 74 | |
| BC027915.1 | - | 3 - 4 | 302 | 5223298 | 74 | |
| M94547.1 | - | 3 - 4 | 306 | 74 | ||
| W17098.1 | - | 4 - 5 | 306 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001336.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07020727.hg.1 | ENST00000223364.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07020727.hg.1 | ENST00000434895.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07020727.hg.1 | ENST00000458240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07020727.hg.1 | NM_021223.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07020727.hg.1 | TC07001336.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000223364.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000223364.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000223364.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000223364.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000223364.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000223364.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000223364.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000223364.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000223364.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000431007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000431007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000431007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000431007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000431007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000431007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000431289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000431289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000431289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000431289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000431289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000431289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000434895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000434895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000434895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000434895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000434895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000434895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000434895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000434895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000434895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000446581.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000446581.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000446581.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000446581.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000446581.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000446581.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000447951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000447951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000447951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000447951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000447951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000447951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000447951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000447951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000457314.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000457314.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000457314.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000457314.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000457314.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000457314.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000457314.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000457314.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000457314.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000457314.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000457314.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000457910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000457910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000457910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000457910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000457910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000458240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000458240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000458240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000458240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000458240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000458240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000458240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000458240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000458240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000458240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000476118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000476118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000476118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000476118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000476118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | ENST00000476118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | NM_021223.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | NM_021223.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | NM_021223.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | NM_021223.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | NM_021223.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | NM_021223.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | NM_021223.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | NM_021223.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010912.hg.1 | NM_021223.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000223364.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000223364.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000223364.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000223364.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000223364.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000223364.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000223364.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000223364.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000223364.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000431007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000431007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000431007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000431007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000431007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000431007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000431289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000431289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000431289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000431289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000431289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000431289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000434895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000434895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000434895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000434895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000434895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000434895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000434895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000434895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000434895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000446581.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000446581.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000446581.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000446581.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000446581.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000446581.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000447951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000447951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000447951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000447951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000447951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000447951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000447951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000447951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000457314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000457314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000457314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000457314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000457314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000457314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000457314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000457314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000457314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000457314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000457314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000457910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000457910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000457910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000457910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000457910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000458240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000458240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000458240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000458240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000458240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000458240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000458240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000458240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000458240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000458240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000476118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000476118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000476118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000476118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000476118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | ENST00000476118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | NM_021223.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | NM_021223.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | NM_021223.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | NM_021223.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | NM_021223.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | NM_021223.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | NM_021223.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | NM_021223.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010912.hg.1 | NM_021223.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700177783.hg.1 | ENST00000223364.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700177783.hg.1 | ENST00000431289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700177783.hg.1 | ENST00000434895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700177783.hg.1 | ENST00000446581.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700177783.hg.1 | ENST00000447951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700177783.hg.1 | ENST00000457314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700177783.hg.1 | ENST00000458240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700177783.hg.1 | ENST00000476118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700177783.hg.1 | NM_021223.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700177783.hg.1 | ENST00000223364.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700177783.hg.1 | ENST00000431289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700177783.hg.1 | ENST00000434895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700177783.hg.1 | ENST00000446581.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700177783.hg.1 | ENST00000447951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700177783.hg.1 | ENST00000457314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700177783.hg.1 | ENST00000458240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700177783.hg.1 | ENST00000476118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700177783.hg.1 | NM_021223.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700177783.hg.1 | TC0700010912.hg.1 | Clariom D TC |

MYL7
Genomic Map