| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010936.hg.1 |
ENST00000256103.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010936.hg.1 |
ENST00000256103.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010936.hg.1 |
ENST00000256103.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010936.hg.1 |
ENST00000256103.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010936.hg.1 |
ENST00000256103.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010936.hg.1 |
ENST00000256103.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010936.hg.1 |
ENST00000256103.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010936.hg.1 |
ENST00000256103.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010936.hg.1 |
ENST00000256103.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010936.hg.1 |
ENST00000519260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010936.hg.1 |
ENST00000519260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010936.hg.1 |
ENST00000519260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010936.hg.1 |
ENST00000519260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010936.hg.1 |
ENST00000519260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010936.hg.1 |
NM_002677.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010936.hg.1 |
NM_002677.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010936.hg.1 |
NM_002677.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010936.hg.1 |
NM_002677.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010936.hg.1 |
NM_002677.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010936.hg.1 |
NM_002677.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010936.hg.1 |
NM_002677.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010936.hg.1 |
NM_002677.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08017588.hg.1 |
ENST00000256103.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08017588.hg.1 |
ENST00000519260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08017588.hg.1 |
NM_002677.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08017588.hg.1 |
TC08001363.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0800010936.hg.1 |
ENST00000256103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010936.hg.1 |
ENST00000256103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010936.hg.1 |
ENST00000256103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010936.hg.1 |
ENST00000256103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010936.hg.1 |
ENST00000256103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010936.hg.1 |
ENST00000256103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010936.hg.1 |
ENST00000256103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010936.hg.1 |
ENST00000256103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010936.hg.1 |
ENST00000256103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010936.hg.1 |
ENST00000519260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010936.hg.1 |
ENST00000519260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010936.hg.1 |
ENST00000519260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010936.hg.1 |
ENST00000519260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010936.hg.1 |
ENST00000519260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010936.hg.1 |
NM_002677.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010936.hg.1 |
NM_002677.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010936.hg.1 |
NM_002677.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010936.hg.1 |
NM_002677.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010936.hg.1 |
NM_002677.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010936.hg.1 |
NM_002677.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010936.hg.1 |
NM_002677.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010936.hg.1 |
NM_002677.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800174262.hg.1 |
ENST00000256103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800174262.hg.1 |
ENST00000519260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800174262.hg.1 |
NM_002677.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800174262.hg.1 |
ENST00000256103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800174262.hg.1 |
ENST00000519260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800174262.hg.1 |
NM_002677.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800174262.hg.1 |
TC0800010936.hg.1 |
Clariom D TC |