| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19000192.hg.1 |
ENST00000234371.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19000192.hg.1 |
NM_032551.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19000192.hg.1 |
TC19000022.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
ENST00000234371.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
ENST00000234371.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
ENST00000234371.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
ENST00000234371.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
ENST00000234371.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
ENST00000234371.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
ENST00000234371.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
ENST00000234371.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
ENST00000234371.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
ENST00000234371.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
ENST00000592648.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
ENST00000592648.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
ENST00000592648.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
ENST00000592648.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
ENST00000592648.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
ENST00000606939.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
ENST00000606939.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
ENST00000606939.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
ENST00000606939.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
ENST00000606939.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
NM_032551.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
NM_032551.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
NM_032551.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
NM_032551.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
NM_032551.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
NM_032551.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
NM_032551.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
NM_032551.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
NM_032551.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006498.hg.1 |
NM_032551.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
ENST00000234371.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
ENST00000234371.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
ENST00000234371.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
ENST00000234371.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
ENST00000234371.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
ENST00000234371.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
ENST00000234371.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
ENST00000234371.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
ENST00000234371.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
ENST00000234371.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
ENST00000592648.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
ENST00000592648.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
ENST00000592648.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
ENST00000592648.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
ENST00000592648.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
ENST00000606939.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
ENST00000606939.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
ENST00000606939.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
ENST00000606939.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
ENST00000606939.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
NM_032551.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
NM_032551.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
NM_032551.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
NM_032551.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
NM_032551.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
NM_032551.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
NM_032551.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
NM_032551.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
NM_032551.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006498.hg.1 |
NM_032551.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900144590.hg.1 |
ENST00000234371.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900144590.hg.1 |
NM_032551.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900144590.hg.1 |
TC1900006498.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900144590.hg.1 |
ENST00000234371.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900144590.hg.1 |
NM_032551.4 |
Refseq |