| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009601.hg.1 |
ENST00000415962.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19006300.hg.1 |
ENST00000246554.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19006300.hg.1 |
ENST00000392201.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19006300.hg.1 |
NM_001863.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19006300.hg.1 |
TC19000479.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
ENST00000246554.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
ENST00000246554.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
ENST00000246554.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
ENST00000246554.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
ENST00000246554.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
ENST00000246554.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
ENST00000246554.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
ENST00000246554.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
ENST00000246554.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
ENST00000392201.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
ENST00000392201.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
ENST00000392201.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
ENST00000392201.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
ENST00000392201.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
ENST00000590618.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
ENST00000590618.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
ENST00000590618.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
ENST00000590618.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
ENST00000592141.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
ENST00000592141.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
ENST00000592141.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
ENST00000592141.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
ENST00000592141.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
ENST00000592141.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
ENST00000592141.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
NM_001863.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
NM_001863.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
NM_001863.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
NM_001863.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
NM_001863.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
NM_001863.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
NM_001863.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007871.hg.1 |
NM_001863.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
ENST00000246554.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
ENST00000246554.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
ENST00000246554.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
ENST00000246554.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
ENST00000246554.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
ENST00000246554.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
ENST00000246554.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
ENST00000246554.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
ENST00000246554.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
ENST00000392201.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
ENST00000392201.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
ENST00000392201.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
ENST00000392201.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
ENST00000392201.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
ENST00000590618.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
ENST00000590618.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
ENST00000590618.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
ENST00000590618.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
ENST00000592141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
ENST00000592141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
ENST00000592141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
ENST00000592141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
ENST00000592141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
ENST00000592141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
ENST00000592141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
NM_001863.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
NM_001863.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
NM_001863.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
NM_001863.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
NM_001863.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
NM_001863.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
NM_001863.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007871.hg.1 |
NM_001863.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900158423.hg.1 |
ENST00000246554.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900158423.hg.1 |
ENST00000392201.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900158423.hg.1 |
ENST00000592141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900158423.hg.1 |
NM_001863.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900158423.hg.1 |
TC1900007871.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900158423.hg.1 |
ENST00000246554.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900158423.hg.1 |
ENST00000392201.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900158423.hg.1 |
ENST00000592141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900158423.hg.1 |
NM_001863.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009601.hg.1 |
ENST00000415962.1 |
Ensembl |