Assay Details
| Gene Symbol: FAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001447.2 | NP_001438.1 | 11 - 12 | 9200 | 80 | |
| XM_006714761.3 | XP_006714824.1 | 12 - 13 | 9771 | 80 | ||
| XM_011537600.2 | XP_011535902.1 | 14 - 15 | 9539 | 80 | ||
| XM_011537603.2 | XP_011535905.1 | 12 - 13 | 9472 | 80 | ||
| XM_017009224.1 | XP_016864713.1 | 14 - 15 | 9541 | 80 | ||
| XM_017009225.1 | XP_016864714.1 | 13 - 14 | 9383 | 80 | ||
| GenBank mRNA | AF231022.1 | - | 11 - 12 | 9200 | 80 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001954.hg.1
Set Membership: |
|

FAT2
Genomic Map