Assay Details
| Gene Symbol: CCDC158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042784.1 | NP_001036249.1 | 18 - 19 | 2961 | 80 | |
| XM_005262974.4 | XP_005263031.1 | 19 - 20 | 3088 | 80 | ||
| GenBank mRNA | AK098600.1 | - | 7 - 8 | 978 | 80 | |
| BC086869.1 | - | 14 - 15 | 2331 | 80 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001309.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017990.hg.1 | ENST00000388914.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017990.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017990.hg.1 | TC04001309.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400175432.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175432.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175432.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400175432.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175432.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400175432.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400175432.hg.1 | TC0400011061.hg.1 | Clariom D TC |

CCDC158
Genomic Map