| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000359942.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000359942.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000359942.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000359942.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000359942.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000359942.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000359942.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000359942.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000359942.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000359942.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000359942.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000359942.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000430643.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000430643.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000461718.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000461718.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000495072.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000495072.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000495072.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
ENST00000495072.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01040770.hg.1 |
ENST00000359942.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01040770.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01040770.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01040770.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01040770.hg.1 |
TC01002615.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000359942.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000359942.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000359942.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000359942.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000359942.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000359942.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000359942.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000359942.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000359942.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000359942.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000359942.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000359942.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000430643.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000430643.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000461718.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000461718.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000495072.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000495072.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000495072.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
ENST00000495072.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014032.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100204948.hg.1 |
ENST00000359942.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100204948.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100204948.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100204948.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100204948.hg.1 |
ENST00000359942.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100204948.hg.1 |
ENST00000396478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100204948.hg.1 |
NM_001145349.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100204948.hg.1 |
NM_005897.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100204948.hg.1 |
TC0100014032.hg.1 |
Clariom D TC |