Assay Details
| Gene Symbol: IPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145349.1 | NP_001138821.1 | 4 - 5 | 1150 | 89 | |
| NM_005897.2 | NP_005888.1 | 4 - 5 | 1150 | 89 | ||
| XM_005270839.4 | XP_005270896.1 | 4 - 5 | 1150 | 89 | ||
| XM_006710623.3 | XP_006710686.1 | 4 - 5 | 1150 | 89 | ||
| XM_011541394.2 | XP_011539696.1 | 4 - 5 | 1150 | 89 | ||
| XM_017001210.1 | XP_016856699.1 | 4 - 5 | 1150 | 89 | ||
| GenBank mRNA | AF156857.1 | - | 3 - 4 | 933 | 89 | |
| AK023880.1 | - | 4 - 5 | 986 | 89 | ||
| BC032544.1 | - | 4 - 5 | 991 | 5535760 | 89 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002615.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01040769.hg.1 | ENST00000359942.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040769.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040769.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040769.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040769.hg.1 | TC01002615.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040768.hg.1 | ENST00000359942.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040768.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040768.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040768.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040768.hg.1 | TC01002615.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000430643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000430643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000461718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000461718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000495072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000495072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000495072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000495072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000430643.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000430643.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000461718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000461718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000495072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000495072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000495072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000495072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204946.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204946.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204946.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204946.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204946.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204946.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204946.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204946.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204946.hg.1 | TC0100014032.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100204947.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204947.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204947.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204947.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204947.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204947.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204947.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204947.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204947.hg.1 | TC0100014032.hg.1 | Clariom D TC |

IPP
Genomic Map