Assay Details
| Gene Symbol: SLC25A20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000387.5 | NP_000378.1 | 1 - 2 | 307 | 88 | |
| GenBank mRNA | AK312962.1 | - | 1 - 2 | 200 | 88 | |
| BC001689.2 | - | 1 - 2 | 179 | 3050073 | 88 | |
| BI913588.1 | - | 1 - 2 | 210 | 88 | ||
| DC360427.1 | - | 1 - 2 | 308 | 88 | ||
| Y10319.1 | - | 1 - 2 | 170 | 88 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001390.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03024520.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024520.hg.1 | ENST00000430379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024520.hg.1 | ENST00000544097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024520.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024520.hg.1 | TC03001390.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008250.hg.1 | ENST00000408043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000479050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000479050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000479050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000479050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000479050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024519.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024519.hg.1 | ENST00000430379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024519.hg.1 | ENST00000544097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024519.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024519.hg.1 | TC03001390.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024522.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024522.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024522.hg.1 | TC03001390.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024517.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024517.hg.1 | ENST00000430379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024517.hg.1 | ENST00000544097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024517.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024517.hg.1 | TC03001390.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0300181336.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181336.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181336.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181336.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300181336.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181336.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181336.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181336.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300181336.hg.1 | TC0300011038.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300181335.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181335.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181335.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181335.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300181335.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181335.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181335.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181335.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300181335.hg.1 | TC0300011038.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300181337.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181337.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181337.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181337.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300181337.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181337.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181337.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181337.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300181337.hg.1 | TC0300011038.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000479050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000479050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000479050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000479050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000479050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008250.hg.1 | ENST00000408043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181333.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181333.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181333.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181333.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300181333.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181333.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181333.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181333.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300181333.hg.1 | TC0300011038.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300181339.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181339.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300181339.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181339.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300181339.hg.1 | TC0300011038.hg.1 | Clariom D TC |

SLC25A20
Genomic Map