Assay Details
| Gene Symbol: RASSF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014737.2 | NP_055552.1 | 2 - 3 | 83 | ||
| XM_005260895.3 | XP_005260952.1 | 2 - 3 | 83 | |||
| XM_011529410.1 | XP_011527712.1 | 2 - 3 | 83 | |||
| XM_011529411.1 | XP_011527713.1 | 2 - 3 | 83 | |||
| GenBank mRNA | AK222650.1 | - | 2 - 3 | 125 | 83 | |
| AK291458.1 | - | 2 - 3 | 117 | 83 | ||
| AY154470.1 | - | 2 - 3 | 162 | 83 | ||
| CR627436.1 | - | 2 - 3 | 182 | 83 | ||
| D79990.1 | - | 2 - 3 | 162 | 83 | ||
| DA333367.1 | - | 2 - 3 | 83 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000587.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK291458.1,AY154470.1,CR627436.1,D79990.1,NM_014737.2,XM_005260895.3,XM_011529410.1,XM_011529411.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
ossification
protein phosphorylation
cell cycle
positive regulation of protein autophosphorylation
negative regulation of peptidyl-serine phosphorylation
epidermal growth factor receptor signaling pathway via I-kappaB kinase/NF-kappaB cascade
positive regulation of apoptotic process
regulation of osteoblast differentiation
regulation of osteoclast differentiation
positive regulation of protein kinase activity
positive regulation of JNK cascade
bone remodeling
homeostasis of number of cells
protein stabilization
negative regulation of NIK/NF-kappaB signaling
protein kinase activity
protein binding
skeletal system development
ossification
protein phosphorylation
cell cycle
positive regulation of protein autophosphorylation
negative regulation of peptidyl-serine phosphorylation
epidermal growth factor receptor signaling pathway via I-kappaB kinase/NF-kappaB cascade
positive regulation of apoptotic process
regulation of osteoblast differentiation
regulation of osteoclast differentiation
positive regulation of protein kinase activity
positive regulation of JNK cascade
bone remodeling
homeostasis of number of cells
protein stabilization
negative regulation of NIK/NF-kappaB signaling
protein kinase activity
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000478553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000478553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000478553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000478553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000478553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000478553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000478553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000478553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000478553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000478553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000478553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000478553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | ENST00000478553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008574.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008574.hg.1 | ENST00000379400.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008574.hg.1 | ENST00000474232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008574.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008574.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008574.hg.1 | TC20000587.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2000158279.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158279.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158279.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000158279.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000158279.hg.1 | TC2000008267.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000158279.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158279.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158279.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000158279.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000478553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000478553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000478553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000478553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000478553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000478553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000478553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000478553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000478553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000478553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000478553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000478553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | ENST00000478553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008267.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000158280.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158280.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158280.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000158280.hg.1 | NM_170774.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000158280.hg.1 | TC2000008267.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000158280.hg.1 | ENST00000379376.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158280.hg.1 | ENST00000379400.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158280.hg.1 | NM_014737.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000158280.hg.1 | NM_170774.1 | Refseq |

RASSF2
Genomic Map