| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000478553.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000478553.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000478553.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000478553.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000478553.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000478553.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000478553.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000478553.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000478553.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000478553.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000478553.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000478553.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
ENST00000478553.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008573.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008573.hg.1 |
ENST00000379400.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008573.hg.1 |
ENST00000474232.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008573.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008573.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008573.hg.1 |
TC20000587.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2000158277.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158277.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158277.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000158277.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000158277.hg.1 |
TC2000008267.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000158277.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158277.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158277.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000158277.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000379400.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000478553.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000478553.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000478553.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000478553.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000478553.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000478553.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000478553.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000478553.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000478553.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000478553.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000478553.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000478553.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
ENST00000478553.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_014737.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008267.hg.1 |
NM_170774.1 |
Refseq |