Assay Details
| Gene Symbol: IQCE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287499.1 | NP_001274428.1 | 17 - 18 | 1811 | 74 | |
| NM_001287500.1 | NP_001274429.1 | 16 - 17 | 1763 | 74 | ||
| NM_001287501.1 | NP_001274430.1 | 15 - 16 | 1717 | 74 | ||
| NM_001287502.1 | NP_001274431.1 | 15 - 16 | 1717 | 74 | ||
| NM_152558.4 | NP_689771.3 | 17 - 18 | 1811 | 74 | ||
| XM_006715676.2 | XP_006715739.1 | 15 - 16 | 1565 | 74 | ||
| XM_011515242.1 | XP_011513544.1 | 16 - 17 | 1670 | 74 | ||
| XM_011515243.1 | XP_011513545.1 | 15 - 16 | 1692 | 74 | ||
| XM_011515244.2 | XP_011513546.1 | 10 - 11 | 1106 | 74 | ||
| XM_017011902.1 | XP_016867391.1 | 17 - 18 | 1873 | 74 | ||
| XM_017011903.1 | XP_016867392.1 | 17 - 18 | 1751 | 74 | ||
| GenBank mRNA | AB028946.2 | - | 15 - 16 | 1514 | 74 | |
| AK092404.1 | - | 17 - 18 | 1705 | 74 | ||
| AK293372.1 | - | 16 - 17 | 1586 | 74 | ||
| AK302053.1 | - | 15 - 16 | 1665 | 74 | ||
| AL136792.1 | - | 16 - 17 | 1492 | 74 | ||
| BC043150.1 | - | 15 - 16 | 1637 | 5297871 | 74 | |
| BC071858.1 | - | 17 - 18 | 1785 | 5478116 | 74 | |
| KJ534872.1 | - | 17 - 18 | 1601 | 74 | ||
| KJ535081.1 | - | 15 - 16 | 1507 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

IQCE
Genomic Map