Assay Details
| Gene Symbol: BRINP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014618.2 | NP_055433.2 | 6 - 7 | 1376 | 67 | |
| GenBank mRNA | AF027734.1 | - | 6 - 7 | 1338 | 67 | |
| BC065196.1 | - | 6 - 7 | 1329 | 5245480 | 67 | |
| BC071702.1 | - | 2 - 3 | 324 | 5244680 | 67 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001533.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09019134.hg.1 | ENST00000265922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019134.hg.1 | ENST00000373964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019134.hg.1 | ENST00000373969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019134.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019134.hg.1 | TC09001533.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019132.hg.1 | ENST00000265922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019132.hg.1 | ENST00000373969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019132.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019132.hg.1 | TC09001533.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | ENST00000373964.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | ENST00000373964.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | ENST00000373964.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | ENST00000373964.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | ENST00000373964.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | ENST00000373964.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | ENST00000373964.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | ENST00000373964.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | ENST00000373964.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | ENST00000373964.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | ENST00000482797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | ENST00000482797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019131.hg.1 | ENST00000265922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019131.hg.1 | ENST00000373969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019131.hg.1 | ENST00000482797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019131.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019131.hg.1 | TC09001533.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0900177384.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900177384.hg.1 | ENST00000482797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900177384.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900177384.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900177384.hg.1 | ENST00000482797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900177384.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900177384.hg.1 | TC0900011370.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | ENST00000373964.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | ENST00000373964.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | ENST00000373964.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | ENST00000373964.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | ENST00000373964.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | ENST00000373964.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | ENST00000373964.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | ENST00000373964.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | ENST00000373964.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | ENST00000373964.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | ENST00000482797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | ENST00000482797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011370.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900177385.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900177385.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900177385.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900177385.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900177385.hg.1 | TC0900011370.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900177387.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900177387.hg.1 | ENST00000373964.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900177387.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900177387.hg.1 | ENST00000265922.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900177387.hg.1 | ENST00000373964.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900177387.hg.1 | NM_014618.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900177387.hg.1 | TC0900011370.hg.1 | Clariom D TC |

BRINP1
Genomic Map