Assay Details
| Gene Symbol: SLC27A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017372.2 | NP_001017372.1 | 3 - 4 | 1858 | 150 | |
| NM_001317984.1 | NP_001304913.1 | 4 - 5 | 1682 | 150 | ||
| NM_014031.4 | NP_054750.1 | 4 - 5 | 1633 | 150 | ||
| GenBank mRNA | AF064254.1 | - | 3 - 4 | 1000 | 150 | |
| AK313456.1 | - | 4 - 5 | 1281 | 150 | ||
| AL833117.1 | - | 3 - 4 | 1431 | 150 | ||
| BC041945.1 | - | 3 - 4 | 1857 | 5301306 | 150 | |
| BX537383.1 | - | 4 - 5 | 1243 | 150 | ||
| CR457129.1 | - | 3 - 4 | 847 | 150 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000613.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05008557.hg.1 | ENST00000262462.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05008557.hg.1 | ENST00000395266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05008557.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05008557.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05008557.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05008557.hg.1 | TC05000613.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05008558.hg.1 | ENST00000262462.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05008558.hg.1 | ENST00000395266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05008558.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05008558.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05008558.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05008558.hg.1 | TC05000613.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500194032.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500194032.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500194032.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500194032.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500194032.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500194032.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500194032.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500194032.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500194032.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500194032.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500194032.hg.1 | TC0500013224.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013224.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500194031.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500194031.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500194031.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500194031.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500194031.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500194031.hg.1 | ENST00000262462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500194031.hg.1 | ENST00000395266.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500194031.hg.1 | ENST00000506176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500194031.hg.1 | NM_001017372.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500194031.hg.1 | NM_014031.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500194031.hg.1 | TC0500013224.hg.1 | Clariom D TC |

SLC27A6
Genomic Map