Assay Details
| Gene Symbol: CRYZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130042.1 | NP_001123514.1 | 2 - 3 | 495 | 146 | |
| XM_005270491.4 | XP_005270548.1 | 2 - 3 | 133 | 146 | ||
| XM_017000367.1 | XP_016855856.1 | 2 - 3 | 130 | 146 | ||
| GenBank mRNA | AB209714.1 | - | 2 - 3 | 127 | 146 | |
| BC070058.1 | - | 2 - 3 | 137 | 5267616 | 146 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC070058.1, NM_001130042.1, XM_005270491.4, XM_017000367.1 transcripts.
Set Membership: |
|

CRYZ
Genomic Map