Assay Details
| Gene Symbol: EML2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193268.1 | NP_001180197.1 | 5 - 6 | 720 | 88 | |
| NM_001193269.1 | NP_001180198.1 | 5 - 6 | 624 | 88 | ||
| XM_006723125.2 | XP_006723188.1 | 5 - 6 | 801 | 88 | ||
| XM_006723126.2 | XP_006723189.1 | 5 - 6 | 609 | 88 | ||
| XM_011526694.2 | XP_011524996.1 | 5 - 6 | 804 | 88 | ||
| XM_011526695.2 | XP_011524997.1 | 5 - 6 | 804 | 88 | ||
| XM_017026551.1 | XP_016882040.1 | 5 - 6 | 804 | 88 | ||
| GenBank mRNA | AK294668.1 | - | 5 - 6 | 671 | 88 | |
| AK295905.1 | - | 5 - 6 | 624 | 88 | ||
| AK296258.1 | - | 5 - 6 | 553 | 88 | ||
| AK302961.1 | - | 5 - 6 | 684 | 88 | ||
More Information
Set Membership: |
|

EML2
Genomic Map