Assay Details
| Gene Symbol: NUP210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024923.3 | NP_079199.2 | 29 - 30 | 4031 | 105 | |
| XM_017006018.1 | XP_016861507.1 | 11 - 12 | 1864 | 105 | ||
| XM_017006019.1 | XP_016861508.1 | 11 - 12 | 1831 | 105 | ||
| XM_017006020.1 | XP_016861509.1 | 11 - 12 | 3903 | 105 | ||
| GenBank mRNA | AB020713.1 | - | 9 - 10 | 1047 | 105 | |
| AK074101.1 | - | 14 - 15 | 1653 | 105 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001188.hg.1
Set Membership: |
|

NUP210
Genomic Map