| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000572078.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000572078.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000572078.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000572078.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000572078.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000575185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000575185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000575185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000575185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000575796.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000575796.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000575796.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000575796.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000576069.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000576069.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000576069.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000576069.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000576069.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000576069.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
ENST00000576069.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17000418.hg.1 |
ENST00000333476.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17000418.hg.1 |
ENST00000355530.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17000418.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17000418.hg.1 |
TC17000041.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1700145246.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700145246.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700145246.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700145246.hg.1 |
TC1700006626.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700145246.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700145246.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700145246.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000355530.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000572078.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000572078.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000572078.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000572078.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000572078.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000575185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000575185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000575185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000575185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000575194.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000575796.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000575796.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000575796.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000575796.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000576069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000576069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000576069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000576069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000576069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000576069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
ENST00000576069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006626.hg.1 |
NM_182538.4 |
Refseq |