Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | melanoma associated antigen (mutated) 1 | MUM1 |
EXPAND1, HSPC211, MUM-1 |
-8.4kb | Anti-sense | Chr. 19 - 1354977 - 1378431 on NCBI Build GRCh38 |
| Downstream | guanidinoacetate N-methyltransferase | GAMT |
CCDS2, HEL-S-20, PIG2, TP53I2 |
8.7kb | Sense | Chr. 19 - 1397026 - 1401570 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map