Assay Details
| Gene Symbol: KLK13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY923172.1 | - | 4 - 5 | 905 | 115 | |
| AY923174.1 | - | 3 - 4 | 821 | 115 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000156476.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000156476.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000156476.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000156476.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000156476.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000156476.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000156476.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000156476.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000335422.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000335422.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000335422.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000376799.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000376799.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000376799.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000376799.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000376799.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000376799.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000376799.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000441527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000441527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000441527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000441527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000441527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000441527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000441527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000441527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000441527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000441527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000441527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000595547.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000595547.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000595547.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000595547.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000595547.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000595793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000595793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000595793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000595793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000595793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000595793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000595793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000595793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000596955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000596955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000596955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000596955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000596955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000596955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000596955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000601975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000601975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000601975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000601975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000601975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000601975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000601975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000601975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000601975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000602090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000602090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000602090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000602090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000602090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000602090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000602090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000602090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | ENST00000602090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | NM_015596.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | NM_015596.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | NM_015596.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | NM_015596.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | NM_015596.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | NM_015596.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | NM_015596.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | NM_015596.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011236.hg.1 | NM_015596.2 | Refseq |

KLK13
Genomic Map