Assay Details
| Gene Symbol: GKAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135953.1 | NP_001129425.1 | 6 - 7 | 1057 | 84 | |
| GenBank mRNA | AB033132.1 | - | 6 - 7 | 853 | 84 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000462223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000462223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000485742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000485742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000485742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000485742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000491634.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000491634.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | ENST00000491634.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376365.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000376371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000388782.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000462223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000462223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000485742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000485742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000485742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000485742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000491634.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000491634.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | ENST00000491634.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_001135953.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_025211.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010580.hg.1 | NM_025211.3 | Refseq |

GKAP1
Genomic Map