| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X018305.hg.1 |
ENST00000361420.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X018305.hg.1 |
ENST00000370903.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X018305.hg.1 |
ENST00000370904.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X018305.hg.1 |
ENST00000370910.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X018305.hg.1 |
NM_001170961.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X018305.hg.1 |
NM_001170962.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X018305.hg.1 |
NM_001555.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X018305.hg.1 |
TC0X001365.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0X00173434.hg.1 |
ENST00000361420.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00173434.hg.1 |
ENST00000370903.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00173434.hg.1 |
ENST00000370904.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00173434.hg.1 |
ENST00000370910.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00173434.hg.1 |
NM_001170961.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00173434.hg.1 |
NM_001170962.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00173434.hg.1 |
NM_001555.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00173434.hg.1 |
ENST00000361420.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00173434.hg.1 |
ENST00000370903.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00173434.hg.1 |
ENST00000370904.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00173434.hg.1 |
ENST00000370910.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00173434.hg.1 |
NM_001170961.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00173434.hg.1 |
NM_001170962.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00173434.hg.1 |
NM_001555.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00173434.hg.1 |
TC0X00010818.hg.1 |
Clariom D TC |