Assay Details
| Gene Symbol: RASSF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005260895.3 | XP_005260952.1 | 1 - 2 | 156 | 107 | |
| XM_011529411.1 | XP_011527713.1 | 1 - 2 | 156 | 107 | ||
| XM_017028152.1 | XP_016883641.1 | 1 - 2 | 151 | 107 | ||
| GenBank mRNA | CR627436.1 | - | 1 - 2 | 110 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the CR627436.1, XM_005260895.3, XM_011529411.1, XM_017028152.1 transcripts.
Set Membership: |
|

RASSF2
Genomic Map