Assay Details
| Gene Symbol: TMEM30A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018247.3 | NP_060717.1 | 1 - 2 | 511 | 91 | |
| GenBank mRNA | AK001718.1 | - | 1 - 2 | 380 | 91 | |
| AK295567.1 | - | 2 - 3 | 332 | 91 | ||
| AL832815.1 | - | 1 - 2 | 373 | 91 | ||
| DB239737.1 | - | 1 - 2 | 475 | 91 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001881.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06022932.hg.1 | ENST00000230461.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022932.hg.1 | ENST00000370050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022932.hg.1 | ENST00000545449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022932.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022932.hg.1 | TC06001881.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000518161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000518161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000518161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000518161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000518161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022935.hg.1 | ENST00000230461.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022935.hg.1 | ENST00000475111.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022935.hg.1 | ENST00000545449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022935.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022935.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022935.hg.1 | TC06001881.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022939.hg.1 | ENST00000230461.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022939.hg.1 | ENST00000475111.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022939.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022939.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022939.hg.1 | TC06001881.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022936.hg.1 | ENST00000230461.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022936.hg.1 | ENST00000475111.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022936.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022936.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022936.hg.1 | TC06001881.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022938.hg.1 | ENST00000230461.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022938.hg.1 | ENST00000475111.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022938.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022938.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022938.hg.1 | TC06001881.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000518161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000518161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000518161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000518161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000518161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600184984.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600184984.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600184984.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600184984.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600184984.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600184984.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600184984.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600184984.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600184984.hg.1 | TC0600012316.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600184985.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600184985.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600184985.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600184985.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600184985.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600184985.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600184985.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600184985.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600184985.hg.1 | TC0600012316.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600184979.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600184979.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600184979.hg.1 | ENST00000518161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600184979.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600184979.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600184979.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600184979.hg.1 | ENST00000518161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600184979.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600184979.hg.1 | TC0600012316.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600184986.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600184986.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600184986.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600184986.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600184986.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600184986.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600184986.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600184986.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600184986.hg.1 | TC0600012316.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 223984 | - | - |

TMEM30A
Genomic Map