Assay Details
| Gene Symbol: RAPSN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032645.4 | NP_116034.2 | 4 - 5 | 1004 | 72 | |
| GenBank mRNA | AK310002.1 | - | 4 - 5 | 962 | 72 | |
| BC004196.2 | - | 4 - 5 | 969 | 3530551 | 72 | |
| KU178300.1 | - | 4 - 5 | 790 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
synaptic transmission, cholinergic
positive regulation of neuron apoptotic process
skeletal muscle acetylcholine-gated channel clustering
positive regulation of neuromuscular synaptic transmission
regulation of postsynaptic membrane organization
zinc ion binding
acetylcholine receptor binding
ionotropic glutamate receptor binding
protein anchor
chemical synaptic transmission
synaptic transmission, cholinergic
positive regulation of neuron apoptotic process
skeletal muscle acetylcholine-gated channel clustering
positive regulation of neuromuscular synaptic transmission
regulation of postsynaptic membrane organization
zinc ion binding
acetylcholine receptor binding
ionotropic glutamate receptor binding
protein anchor
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000298854.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000298854.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000298854.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000298854.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000298854.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000298854.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000298854.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000298854.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000298854.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000298854.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000298854.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000352508.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000352508.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000352508.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000352508.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000352508.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000352508.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000352508.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000352508.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000524487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000524487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000524487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000524487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000524487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000524487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000524487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000524487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000524487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000528356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000528356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000528356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000529341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000529341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000529341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000529341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000529341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000529341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | ENST00000529341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | NM_005055.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | NM_005055.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | NM_005055.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | NM_005055.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | NM_005055.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | NM_005055.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | NM_005055.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | NM_005055.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | NM_005055.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | NM_005055.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | NM_005055.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | NM_032645.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | NM_032645.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | NM_032645.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | NM_032645.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | NM_032645.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | NM_032645.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | NM_032645.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | NM_032645.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010739.hg.1 | NM_032645.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100181423.hg.1 | ENST00000298854.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181423.hg.1 | ENST00000352508.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181423.hg.1 | ENST00000524487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181423.hg.1 | ENST00000529341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181423.hg.1 | NM_005055.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100181423.hg.1 | NM_032645.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100181423.hg.1 | ENST00000298854.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181423.hg.1 | ENST00000352508.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181423.hg.1 | ENST00000524487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181423.hg.1 | ENST00000529341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181423.hg.1 | NM_005055.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100181423.hg.1 | NM_032645.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100181423.hg.1 | TC1100010739.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000298854.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000298854.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000298854.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000298854.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000298854.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000298854.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000298854.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000298854.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000298854.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000298854.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000298854.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000352508.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000352508.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000352508.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000352508.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000352508.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000352508.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000352508.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000352508.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000524487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000524487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000524487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000524487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000524487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000524487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000524487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000524487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000524487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000528356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000528356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000528356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000529341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000529341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000529341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000529341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000529341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000529341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | ENST00000529341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | NM_005055.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | NM_005055.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | NM_005055.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | NM_005055.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | NM_005055.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | NM_005055.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | NM_005055.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | NM_005055.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | NM_005055.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | NM_005055.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | NM_005055.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | NM_032645.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | NM_032645.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | NM_032645.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | NM_032645.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | NM_032645.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | NM_032645.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | NM_032645.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | NM_032645.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010739.hg.1 | NM_032645.4 | Refseq |

RAPSN
Genomic Map