Assay Details
| Gene Symbol: GNPDA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138335.2 | NP_612208.1 | 3 - 4 | 418 | 148 | |
| GenBank mRNA | AB209904.1 | - | 3 - 4 | 329 | 148 | |
| AF247786.1 | - | 3 - 4 | 379 | 148 | ||
| AK055639.1 | - | 3 - 4 | 338 | 148 | ||
| BC015532.2 | - | 2 - 3 | 249 | 3914747 | 148 | |
| BP274548.1 | - | 3 - 4 | 418 | 148 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001157.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000295448.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000295448.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000295448.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000295448.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000295448.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000295448.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000295448.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000295448.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000295448.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000295448.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000295448.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000295448.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000295448.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000507534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000507534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000507534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000507534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000507534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000507534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000507534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000507534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000507534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000507917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000507917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000507917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000507917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000507917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000507917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000507917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000507917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000507917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000507917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000507917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000507917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000507917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000509756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000509756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000509756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000509756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000509756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000509756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000509756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000509756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000509756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000509756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000509756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000511187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000511187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000511187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000511187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000511187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000608855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000608855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000608855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000608855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000609092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000609092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000609092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000609092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | ENST00000609092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270881.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270881.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270881.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270881.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270881.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270881.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270881.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270881.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270881.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270881.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270881.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270881.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270881.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_001270881.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_138335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_138335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_138335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_138335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_138335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_138335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_138335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_138335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_138335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_138335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_138335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_138335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_138335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_138335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_138335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NM_138335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NR_073094.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NR_073094.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NR_073094.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NR_073094.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NR_073094.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NR_073094.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NR_073094.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NR_073094.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NR_073094.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NR_073094.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NR_073094.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NR_073094.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NR_073094.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NR_073094.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NR_073094.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NR_073095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NR_073095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NR_073095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NR_073095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NR_073095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NR_073095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NR_073095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NR_073095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NR_073095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NR_073095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NR_073095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010580.hg.1 | NR_073095.1 | Refseq |

GNPDA2
Genomic Map