| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020189.hg.1 |
ENST00000358582.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020189.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020189.hg.1 |
TC19001487.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000585624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000585624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000585624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000585624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000593040.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000593040.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000593040.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000593040.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000593040.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020190.hg.1 |
ENST00000358582.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020190.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020190.hg.1 |
TC19001487.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020188.hg.1 |
ENST00000358582.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020188.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020188.hg.1 |
TC19001487.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000585624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000585624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000585624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000585624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000593040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000593040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000593040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000593040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000593040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900205882.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900205882.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900205882.hg.1 |
ENST00000593040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900205882.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900205882.hg.1 |
TC1900011936.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900205882.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900205882.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900205882.hg.1 |
ENST00000593040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900205882.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900205880.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900205880.hg.1 |
ENST00000585624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900205880.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900205880.hg.1 |
ENST00000593040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900205880.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900205880.hg.1 |
TC1900011936.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900205880.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900205880.hg.1 |
ENST00000585624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900205880.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900205880.hg.1 |
ENST00000593040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900205880.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900205879.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900205879.hg.1 |
ENST00000585624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900205879.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900205879.hg.1 |
ENST00000593040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900205879.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900205879.hg.1 |
TC1900011936.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900205879.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900205879.hg.1 |
ENST00000585624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900205879.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900205879.hg.1 |
ENST00000593040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900205879.hg.1 |
NM_152605.3 |
Refseq |