Assay Details
| Gene Symbol: DNAJC22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304944.1 | NP_001291873.1 | 1 - 2 | 631 | 98 | |
| GenBank mRNA | AK055747.1 | - | 1 - 2 | 631 | 98 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK055747.1,NM_001304944.1 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | ENST00000395069.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | ENST00000395069.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | ENST00000395069.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | ENST00000395069.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | ENST00000395069.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | ENST00000395069.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | ENST00000395069.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | ENST00000395069.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | ENST00000551153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | ENST00000551153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | ENST00000551153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | ENST00000551153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | ENST00000551153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | ENST00000552651.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | ENST00000552651.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | NM_024902.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | NM_024902.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | NM_024902.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | NM_024902.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | NM_024902.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | NM_024902.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | NM_024902.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | NM_024902.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | NM_024902.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007560.hg.1 | NM_024902.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | ENST00000395069.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | ENST00000395069.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | ENST00000395069.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | ENST00000395069.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | ENST00000395069.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | ENST00000395069.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | ENST00000395069.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | ENST00000395069.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | ENST00000551153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | ENST00000551153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | ENST00000551153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | ENST00000551153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | ENST00000551153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | ENST00000552651.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | ENST00000552651.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | NM_024902.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | NM_024902.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | NM_024902.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | NM_024902.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | NM_024902.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | NM_024902.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | NM_024902.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | NM_024902.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | NM_024902.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007560.hg.1 | NM_024902.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153229.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153229.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153229.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153229.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153229.hg.1 | TC1200007560.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200153226.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153226.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153226.hg.1 | NM_024902.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153226.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153226.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153226.hg.1 | NM_024902.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153226.hg.1 | TC1200007560.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200153227.hg.1 | ENST00000395069.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153227.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153227.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153227.hg.1 | NM_024902.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153227.hg.1 | ENST00000395069.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153227.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153227.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153227.hg.1 | NM_024902.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153227.hg.1 | TC1200007560.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200153228.hg.1 | ENST00000395069.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153228.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153228.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153228.hg.1 | NM_024902.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153228.hg.1 | ENST00000395069.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153228.hg.1 | ENST00000549441.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153228.hg.1 | NM_001304944.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153228.hg.1 | NM_024902.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153228.hg.1 | TC1200007560.hg.1 | Clariom D TC |

DNAJC22
Genomic Map