Assay Details
| Gene Symbol: DNAJC22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304944.1 | NP_001291873.1 | 2 - 3 | 1096 | 91 | |
| XM_005269155.2 | XP_005269212.1 | 1 - 2 | 145 | 91 | ||
| XM_005269156.2 | XP_005269213.1 | 1 - 2 | 145 | 91 | ||
| XM_005269157.2 | XP_005269214.1 | 1 - 2 | 145 | 91 | ||
| GenBank mRNA | AK055747.1 | - | 2 - 3 | 1096 | 91 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK055747.1,NM_001304944.1,XM_005269155.2,XM_005269156.2,XM_005269157.2 transcripts.
Set Membership: |
|

DNAJC22
Genomic Map