Assay Details
| Gene Symbol: LMBR1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300751.1 | NP_001287680.1 | 13 - 14 | 1368 | 66 | |
| XM_017019627.1 | XP_016875116.1 | 14 - 15 | 1559 | 66 | ||
| XM_017019629.1 | XP_016875118.1 | 14 - 15 | 2414 | 66 | ||
| XM_017019631.1 | XP_016875120.1 | 12 - 13 | 1118 | 66 | ||
| GenBank mRNA | AK001651.1 | - | 13 - 14 | 1318 | 66 | |
| BC008337.1 | - | 13 - 14 | 1298 | 3506347 | 66 | |
| KU178714.1 | - | 13 - 14 | 1023 | 66 | ||
More Information
Set Membership: |
|

LMBR1L
Genomic Map