| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07010539.hg.1 |
ENST00000257745.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07010539.hg.1 |
ENST00000311117.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07010539.hg.1 |
ENST00000334877.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07010539.hg.1 |
ENST00000334914.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07010539.hg.1 |
ENST00000351043.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07010539.hg.1 |
ENST00000393656.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07010539.hg.1 |
ENST00000476671.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07010539.hg.1 |
ENST00000537308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07010539.hg.1 |
NM_018682.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07010539.hg.1 |
NM_182931.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07010539.hg.1 |
TC07000679.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0700161429.hg.1 |
ENST00000257745.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700161429.hg.1 |
ENST00000311117.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700161429.hg.1 |
ENST00000334884.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700161429.hg.1 |
ENST00000334914.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700161429.hg.1 |
ENST00000476671.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700161429.hg.1 |
ENST00000478990.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700161429.hg.1 |
ENST00000482560.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700161429.hg.1 |
ENST00000622386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700161429.hg.1 |
NM_018682.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700161429.hg.1 |
NM_182931.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700161429.hg.1 |
ENST00000257745.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700161429.hg.1 |
ENST00000311117.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700161429.hg.1 |
ENST00000334884.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700161429.hg.1 |
ENST00000334914.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700161429.hg.1 |
ENST00000476671.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700161429.hg.1 |
ENST00000478990.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700161429.hg.1 |
ENST00000482560.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700161429.hg.1 |
ENST00000622386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700161429.hg.1 |
NM_018682.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700161429.hg.1 |
NM_182931.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700161429.hg.1 |
TC0700008690.hg.1 |
Clariom D TC |