Assay Details
| Gene Symbol: METTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005371.5 | NP_005362.3 | 2 - 3 | 322 | 95 | |
| XM_005268873.2 | XP_005268930.1 | 3 - 4 | 422 | 95 | ||
| XM_017019305.1 | XP_016874794.1 | 2 - 3 | 440 | 95 | ||
| GenBank mRNA | AK314851.1 | - | 2 - 3 | 296 | 95 | |
| BC000550.1 | - | 2 - 3 | 293 | 3163932 | 95 | |
| X96698.1 | - | 2 - 3 | 293 | 95 | ||
| Y18643.1 | - | 2 - 3 | 322 | 95 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001635.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000547653.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000547653.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000547653.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000547653.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000547653.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000548504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000548504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000548504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000548504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000548681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000548681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000548681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000548681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000549773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000549773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000549773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000549773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000551117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000551117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000551117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000551117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000551117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000553125.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000553125.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000553125.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000553125.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000553125.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000553125.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12022124.hg.1 | ENST00000324871.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12022124.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12022124.hg.1 | TC12001635.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009274.hg.1 | ENST00000433306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009274.hg.1 | ENST00000433306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009274.hg.1 | ENST00000433306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009274.hg.1 | ENST00000433306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203966.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203966.hg.1 | ENST00000551117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203966.hg.1 | ENST00000553125.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203966.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200203966.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203966.hg.1 | ENST00000551117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203966.hg.1 | ENST00000553125.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203966.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200203966.hg.1 | TC1200012815.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000547653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000547653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000547653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000547653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000547653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000548504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000548504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000548504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000548504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000548681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000548681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000548681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000548681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000549773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000549773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000549773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000549773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000551117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000551117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000551117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000551117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000551117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000553125.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000553125.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000553125.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000553125.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000553125.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000553125.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009274.hg.1 | ENST00000433306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009274.hg.1 | ENST00000433306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009274.hg.1 | ENST00000433306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009274.hg.1 | ENST00000433306.1 | Ensembl |

METTL1
Genomic Map