| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012270.hg.1 |
ENST00000372262.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012270.hg.1 |
ENST00000537909.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012270.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012270.hg.1 |
TC20000898.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000474438.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000474438.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000474438.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000474438.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000474438.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000474438.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000164850.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164850.hg.1 |
ENST00000474438.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164850.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164850.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000164850.hg.1 |
TC2000009263.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000164850.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164850.hg.1 |
ENST00000474438.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164850.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164850.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000474438.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000474438.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000474438.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000474438.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000474438.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000474438.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |