Assay Details
| Gene Symbol: SPICE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017005777.1 | XP_016861266.1 | 1 - 2 | 109 | 80 | |
| GenBank mRNA | AK057966.1 | - | 1 - 2 | 73 | 80 | |
| AL832327.2 | - | 1 - 2 | 75 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017005777.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000460832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000460832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000460832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000460832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000460832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000460832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000460832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000467618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000467618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000467618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000467618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000467618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000467618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000467618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000480527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000480527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000480527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000480527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000480527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000480527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000480527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000480527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000480527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000495812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000495812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000495812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000495812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000495812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000495812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000495812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000495812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000496859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000496859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000496859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000496859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | ENST00000496859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000295872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000460832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000460832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000460832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000460832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000460832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000460832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000460832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000467618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000467618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000467618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000467618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000467618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000467618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000467618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000480527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000480527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000480527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000480527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000480527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000480527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000480527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000480527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000480527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000495812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000495812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000495812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000495812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000495812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000495812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000495812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000495812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000496105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000496859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000496859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000496859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000496859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | ENST00000496859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | NM_144718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012023.hg.1 | NM_144718.3 | Refseq |

SPICE1
Genomic Map